missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ 51 Expansion Short Style Glass Shell Vials without Closures
Short-style shell vials are ideal for storing dry products.
$56.48 - $134.63
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
Sterility | Non-sterile |
Disposable | Single-use |
For Use With (Application) | Ideal for storing dry products |
Catalog Number | Mfr. No. | Volume (English) | Volume (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Volume (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity | ||
03-339-30B
|
N/A | 0.5 dr. | 2 mL | 12 mm | 35 mm |
Pack of 144 Each for $56.48
Case of 10 Pack for $564.80
|
|
||
03-339-30C
|
N/A | 1 dr. | 4 mL | 15 mm | 45 mm |
Pack of 144 Each for $59.10
Case of 10 Pack for $591.00
|
|
||
03-339-30E
|
N/A | 2 dr. | 8 mL | 17 mm | 60 mm |
Pack of 144 Each for $83.80
Case of 6 Pack for $502.80
|
|
||
03-339-30F
|
N/A | 3 dr. | 11 mL | 19 mm | 65 mm |
Pack of 144 Each for $90.03
Case of 6 Pack for $540.18
|
|
||
03-339-30G
|
N/A | 4 dr. | 16 mL | 21 mm | 70 mm |
Pack of 144 Each for $102.97
Case of 4 Pack for $411.88
|
|
||
03-339-30J
|
N/A | 8 dr. | 30 mL | 25 mm | 95 mm |
Pack of 144 Each for $134.63
Case of 4 Pack for $538.52
|
|
||
03-339-30A
|
N/A | 0.25 dr. | 1 mL | 9 mm | 30 mm |
Pack of 144 Each for $57.20
Case of 10 Pack for $572.00
|
|
||
03-339-30H
|
N/A | 6 dr. | 24 mL | 23 mm | 85 mm |
Pack of 144 Each for $124.40
Case of 4 Pack for $497.60
|
|
||
Description
Short-style shell vials are ideal for storing dry products.
- Plain top design
- Lab pack quantities
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Clear | |
Non-sterile | |
Ideal for storing dry products |
Borosilicate Glass | |
Single-use | |
144 |