Eppendorf™ Rotor for Centrifuge 5810 / 5810 R
Eppendorf Rotor for Centrifuge 5810/5810 R - precision redefined. High-capacity centrifugation with varied rotor options, a range of capacities and speeds, and versatile buckets to ensure efficient sample processing. Ideal for diverse applications, guaranteeing accuracy and reproducibility.
$3741.56 - $10048.98
Catalog Number | Mfr. No. | Capacity | Max. Speed | Max. RCF | Includes | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Max. Speed | Max. RCF | Includes | Price | Quantity | |||
5820740000
|
Eppendorf™
5820740000 |
4 x 750 mL | 3,900 rpm | 3,214 x g | 750 mL Round Buckets (x4) |
Each for $7,834.73
|
|
|||
5820755008
|
Eppendorf™
5820755008 |
4 x 750 mL | 3,900 rpm | 3,214 x g | Without Buckets |
Each for $4,414.32
|
|
|||
022639905
|
Eppendorf™
022639905 |
4 x 750 mL | 3,900 rpm | 3,214 x g | Round Buckets, Software Upgrade |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
5820754001
|
Eppendorf™
5820754001 |
4 x 750 mL | 3,900 rpm | 3,214 x g | Plate Buckets (x4) |
Each for $7,930.14
|
|
|||
04-987-252
|
Eppendorf™
022638602 |
4 x 500 mL | 4,000 rpm | 3,220 x g | 500 mL Rectangular Buckets (x4) |
Each for $7,275.71
|
|
|||
36-100-7283
|
Eppendorf™
022638807 |
4 x 500 mL | 4,000 rpm | 3,220 x g | MTP/Flex Buckets (x4) |
Each for $8,585.78
|
|
|||
05-400-64
|
Eppendorf™
022638009 |
4 x 250 mL | 4,000 rpm | 3,220 x g | 250 mL Rectangular Buckets (x4) |
Each for $5,160.76
|
|
|||
05-400-80
|
Eppendorf™
022638041 |
4 x 250 mL | 4,000 rpm | 3,220 x g | MTP Buckets (x4) |
Each for $6,193.17
|
|
|||
05414321
|
Eppendorf™
5820710004* |
2 x 4 MTP, 2 x 1 DWP | 4,500 rpm | 3,486 x g | Buckets (x2), Aerosol-tight Caps (x2), Plate Carriers (x2) |
Each for $5,877.27
|
|
|||
05414144
|
Eppendorf™
022638611 |
4 x 500 mL | 4,000 rpm | 3,220 x g | Without buckets |
Each for $3,741.56
|
|
|||
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
Specifications
Rotor | |
Centrifuge 5810 / 5810 R |