missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6,13-Pentacenedione 95.0+%, TCI America™

Supplier:  TCI America P16855G

 View more versions of this product

Catalog No. P16855G


Explore available promotions

Description

Description

Specifications

Specifications

6,13-Pentacenedione
394°C
MFCD00003709
6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa
C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O
308.336
308.34
Crystalline Powder
3029-32-1
C22H12O2
5 g
UFCVADNIXDUEFZ-UHFFFAOYSA-N
pentacene-6,13-dione
76415
≥95.0% (HPLC)

Chemical Identifiers

3029-32-1
308.336
UFCVADNIXDUEFZ-UHFFFAOYSA-N
76415
C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O
C22H12O2
MFCD00003709
6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa
pentacene-6,13-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions