missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
6,13-Pentacenequinone, 99%, Thermo Scientific Chemicals
$211.29 - $211.29
Identifiants chimiques
CAS | 3029-32-1 |
---|---|
Molecular Formula | C22H12O2 |
Molecular Weight (g/mol) | 308.34 |
MDL Number | MFCD00003709 |
InChI Key | UFCVADNIXDUEFZ-UHFFFAOYSA-N |
Synonym | 6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa |
PubChem CID | 76415 |
IUPAC Name | pentacene-6,13-dione |
SMILES | C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
AC364270010
|
Thermo Scientific Chemicals
364270010 |
1 g | Glass bottle |
chaque for $211.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
3029-32-1 | |
308.34 | |
UFCVADNIXDUEFZ-UHFFFAOYSA-N | |
76415 | |
C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O |
C22H12O2 | |
MFCD00003709 | |
6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa | |
pentacene-6,13-dione |
Spécifications
3029-32-1 | |
100.0 | |
Yellow | |
98.5% min. (HPLC) | |
C22H12O2 | |
1 g | |
6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa | |
UFCVADNIXDUEFZ-UHFFFAOYSA-N | |
pentacene-6,13-dione | |
76415 | |
99% | |
6, 13-Pentacenequinone, 99% |
98.5 | |
394.0°C | |
Authentic | |
Glass bottle | |
MFCD00003709 | |
07,I,451 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,benzene,pyridine,dioxane,,dimethylformamide,nitrobenzene | |
C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O | |
308.34 | |
308.34 | |
Powder |
Sécurité et manipulation
GHS Signal Word: Warning
RUO – Research Use Only