missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(2-furyl)nicotinic acid, Thermo Scientific™

Chemical Identifiers

CAS 884507-36-2
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD06410385
InChI Key ALJRNVXLRKHVQF-UHFFFAOYSA-N
Synonym 6-2-furyl nicotinic acid, 6-furan-2-yl nicotinic acid, 6-furan-2-yl pyridine-3-carboxylic acid, 6-2-furyl pyridine-3-carboxylic acid
PubChem CID 19010290
IUPAC Name 6-(furan-2-yl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C(C=C1)C1=CC=CO1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC54901CB
SDS View Documents
Thermo Scientific Chemicals
CC54901CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC54901DA
SDS View Documents
Thermo Scientific Chemicals
CC54901DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC54901DE
SDS View Documents
Thermo Scientific Chemicals
CC54901DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

884507-36-2
189.17
ALJRNVXLRKHVQF-UHFFFAOYSA-N
19010290
OC(=O)C1=CN=C(C=C1)C1=CC=CO1
C10H7NO3
MFCD06410385
6-2-furyl nicotinic acid, 6-furan-2-yl nicotinic acid, 6-furan-2-yl pyridine-3-carboxylic acid, 6-2-furyl pyridine-3-carboxylic acid
6-(furan-2-yl)pyridine-3-carboxylic acid

Specifications

884507-36-2
C10H7NO3
250mg
ALJRNVXLRKHVQF-UHFFFAOYSA-N
6-(furan-2-yl)pyridine-3-carboxylic acid
19010290
97%
97%
MFCD06410385
6-2-furyl nicotinic acid, 6-furan-2-yl nicotinic acid, 6-furan-2-yl pyridine-3-carboxylic acid, 6-2-furyl pyridine-3-carboxylic acid
OC(=O)C1=CN=C(C=C1)C1=CC=CO1
189.17
189.17
6-(2-Furyl)nicotinic acid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'