missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydrobenzo[b]thiophen-4-one, 98%

CAS: 13414-95-4 | C8H8OS | 152.211 g/mol

$178.30 - $755.55

Chemical Identifiers

CAS 13414-95-4
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00005861
InChI Key GJEKNELSXNSYAQ-UHFFFAOYSA-N
Synonym 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone
PubChem CID 83418
IUPAC Name 6,7-dihydro-5H-1-benzothiophen-4-one
SMILES C1CC2=C(C=CS2)C(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6022004
View Documents
Thermo Scientific Chemicals
H6022004
2 g N/A
Only null left
 
AAH6022009
View Documents
Thermo Scientific Chemicals
H6022009
10 g N/A
Only null left
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

13414-95-4
152.211
GJEKNELSXNSYAQ-UHFFFAOYSA-N
83418
C1CC2=C(C=CS2)C(=O)C1
C8H8OS
MFCD00005861
6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone
6,7-dihydro-5H-1-benzothiophen-4-one

Specifications

13414-95-4
120°C
C8H8OS
MFCD00005861
112276
Soluble in chloroform.
C1CC2=C(C=CS2)C(=O)C1
152.211
152.21
6,7-Dihydrobenzo[b]thiophen-4-one
38°C to 40°C
110°C (230°F)
1.5895
2 g
6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone
GJEKNELSXNSYAQ-UHFFFAOYSA-N
6,7-dihydro-5H-1-benzothiophen-4-one
83418
98%
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 236-510-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only