missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6,7-Dimethoxy-3H-quinazolin-4-one 98.0+%, TCI America™
$110.60 - $339.70
Chemical Identifiers
CAS | 13794-72-4 |
---|---|
Molecular Formula | C10H10N2O3 |
Molecular Weight (g/mol) | 206.20 |
MDL Number | MFCD01570147 |
InChI Key | DMSRMHGCZUXCMJ-UHFFFAOYSA-N |
Synonym | 6,7-dimethoxy-3,4-dihydroquinazolin-4-one, 6,7-dimethoxyquinazolin-4 3h-one, 6,7-dimethoxyquinazolin-4-ol, 6,7-dimethoxy-3,4-dihydroquinazoline-4-one, 6,7-dimethoxy-3h-quinazolin-4-one, 6,7-dimethoxy-4 1h-quinazolinone, 6,7-dimethoxy-4 3h-quinazolinone, 6,7-dimethoxyquinazolin-4 1h-one, 6,7-dimethoxy-4-quinazolone, 6,7-dimethoxy-quinazolin-4-ol |
PubChem CID | 1501878 |
IUPAC Name | 6,7-dimethoxy-1,4-dihydroquinazolin-4-one |
SMILES | COC1=C(OC)C=C2C(=O)N=CNC2=C1 |
Chemical Identifiers
13794-72-4 | |
206.20 | |
DMSRMHGCZUXCMJ-UHFFFAOYSA-N | |
1501878 | |
COC1=C(OC)C=C2C(=O)N=CNC2=C1 |
C10H10N2O3 | |
MFCD01570147 | |
6,7-dimethoxy-3,4-dihydroquinazolin-4-one, 6,7-dimethoxyquinazolin-4 3h-one, 6,7-dimethoxyquinazolin-4-ol, 6,7-dimethoxy-3,4-dihydroquinazoline-4-one, 6,7-dimethoxy-3h-quinazolin-4-one, 6,7-dimethoxy-4 1h-quinazolinone, 6,7-dimethoxy-4 3h-quinazolinone, 6,7-dimethoxyquinazolin-4 1h-one, 6,7-dimethoxy-4-quinazolone, 6,7-dimethoxy-quinazolin-4-ol | |
6,7-dimethoxy-1,4-dihydroquinazolin-4-one |
Specifications
13794-72-4 | |
White-Yellow | |
MFCD01570147 | |
6,7-dimethoxy-3,4-dihydroquinazolin-4-one, 6,7-dimethoxyquinazolin-4 3h-one, 6,7-dimethoxyquinazolin-4-ol, 6,7-dimethoxy-3,4-dihydroquinazoline-4-one, 6,7-dimethoxy-3h-quinazolin-4-one, 6,7-dimethoxy-4 1h-quinazolinone, 6,7-dimethoxy-4 3h-quinazolinone, 6,7-dimethoxyquinazolin-4 1h-one, 6,7-dimethoxy-4-quinazolone, 6,7-dimethoxy-quinazolin-4-ol | |
COC1=C(OC)C=C2C(=O)N=CNC2=C1 | |
206.20 | |
206.20 | |
Crystalline Powder |
298°C | |
C10H10N2O3 | |
1 g | |
DMSRMHGCZUXCMJ-UHFFFAOYSA-N | |
6,7-dimethoxy-1,4-dihydroquinazolin-4-one | |
1501878 | |
≥98.0% (HPLC,T) | |
6,7-Dimethoxy-3H-quinazolin-4-one |
Safety and Handling
TSCA : No