missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Amino-2,4-xylenol 98.0+%, TCI America™
$590.13 - $590.13
Chemical Identifiers
CAS | 41458-65-5 |
---|---|
Molecular Formula | C8H11NO |
Molecular Weight (g/mol) | 137.18 |
MDL Number | MFCD00007691 |
InChI Key | GISWNAMJAQRJPC-UHFFFAOYSA-N |
Synonym | 6-Amino-2,4-dimethylphenol, 2-Hydroxy-3,5-dimethylaniline |
PubChem CID | 100530 |
IUPAC Name | 2-amino-4,6-dimethylphenol |
SMILES | CC1=CC(C)=C(O)C(N)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A096225G
|
TCI America
A096225G |
25 g |
Each for $590.13
|
|
|||||
Chemical Identifiers
41458-65-5 | |
137.18 | |
GISWNAMJAQRJPC-UHFFFAOYSA-N | |
100530 | |
CC1=CC(C)=C(O)C(N)=C1 |
C8H11NO | |
MFCD00007691 | |
6-Amino-2,4-dimethylphenol, 2-Hydroxy-3,5-dimethylaniline | |
2-amino-4,6-dimethylphenol |
Specifications
130°C | |
25 g | |
C8H11NO | |
6-Amino-2,4-dimethylphenol, 2-Hydroxy-3,5-dimethylaniline | |
CC1=CC(C)=C(O)C(N)=C1 | |
137.18 | |
137.18 | |
6-Amino-2,4-xylenol |
41458-65-5 | |
Crystal-Powder at 20°C | |
MFCD00007691 | |
GISWNAMJAQRJPC-UHFFFAOYSA-N | |
2-amino-4,6-dimethylphenol | |
100530 | |
≥98.0% (GC,T) |
Safety and Handling
TSCA : Yes