missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Amino-2-naphthoic Acid 97.0+%, TCI America™
$261.24 - $880.62
Chemical Identifiers
CAS | 116668-47-4 |
---|---|
Molecular Formula | C11H9NO2 |
Molecular Weight (g/mol) | 187.198 |
MDL Number | MFCD01861831 |
InChI Key | NZTPZUIIYNYZKT-UHFFFAOYSA-N |
Synonym | 6-amino-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-amino, 6-amino-2-naphtoic acid, 6-amino-2-naphthoic, 6-aminoisonaphthoic acid, 2-amino-6-naphthoic acid, 6-amino-2-carboxynaphthalene, bidd:gt0289, 6-amino-2-naphthoicacid |
PubChem CID | 2733954 |
IUPAC Name | 6-aminonaphthalene-2-carboxylic acid |
SMILES | C1=CC2=C(C=CC(=C2)N)C=C1C(=O)O |
Chemical Identifiers
116668-47-4 | |
187.198 | |
NZTPZUIIYNYZKT-UHFFFAOYSA-N | |
2733954 | |
C1=CC2=C(C=CC(=C2)N)C=C1C(=O)O |
C11H9NO2 | |
MFCD01861831 | |
6-amino-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-amino, 6-amino-2-naphtoic acid, 6-amino-2-naphthoic, 6-aminoisonaphthoic acid, 2-amino-6-naphthoic acid, 6-amino-2-carboxynaphthalene, bidd:gt0289, 6-amino-2-naphthoicacid | |
6-aminonaphthalene-2-carboxylic acid |
Specifications
116668-47-4 | |
White-Yellow | |
MFCD01861831 | |
6-amino-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-amino, 6-amino-2-naphtoic acid, 6-amino-2-naphthoic, 6-aminoisonaphthoic acid, 2-amino-6-naphthoic acid, 6-amino-2-carboxynaphthalene, bidd:gt0289, 6-amino-2-naphthoicacid | |
C1=CC2=C(C=CC(=C2)N)C=C1C(=O)O | |
187.198 | |
187.20 | |
Crystalline Powder |
225°C | |
C11H9NO2 | |
5 g | |
NZTPZUIIYNYZKT-UHFFFAOYSA-N | |
6-aminonaphthalene-2-carboxylic acid | |
2733954 | |
≥97.0% (HPLC,T) | |
6-Amino-2-naphthoic Acid |
Safety and Handling
TSCA : No