missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-pyridinecarboxylic Acid 98.0+%, TCI America™

$87.60 - $367.22

Chemical Identifiers

CAS 23628-31-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00233711
InChI Key NMCKJFCJIHCHIS-UHFFFAOYSA-N
Synonym 6-aminopicolinic acid, 2-aminopyridine-6-carboxylic acid, 6-amino-2-pyridinecarboxylic acid, 6-amino-2-picolinic acid, 6-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-amino, 2-amino-6-pyridinecarboxylic acid, picolinic acid, 6-amino, pubchem7686, acmc-209g4h
PubChem CID 351652
IUPAC Name 6-aminopyridine-2-carboxylic acid
SMILES NC1=CC=CC(=N1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A21881G
View Documents
TCI America
A21881G
1 g
Each for $87.60
Only null left
 
A21885G
View Documents
TCI America
A21885G
5 g
Each for $367.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

23628-31-1
138.13
NMCKJFCJIHCHIS-UHFFFAOYSA-N
351652
NC1=CC=CC(=N1)C(O)=O
C6H6N2O2
MFCD00233711
6-aminopicolinic acid, 2-aminopyridine-6-carboxylic acid, 6-amino-2-pyridinecarboxylic acid, 6-amino-2-picolinic acid, 6-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-amino, 2-amino-6-pyridinecarboxylic acid, picolinic acid, 6-amino, pubchem7686, acmc-209g4h
6-aminopyridine-2-carboxylic acid

Specifications

23628-31-1
White-Yellow
MFCD00233711
6-aminopicolinic acid, 2-aminopyridine-6-carboxylic acid, 6-amino-2-pyridinecarboxylic acid, 6-amino-2-picolinic acid, 6-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-amino, 2-amino-6-pyridinecarboxylic acid, picolinic acid, 6-amino, pubchem7686, acmc-209g4h
NC1=CC=CC(=N1)C(O)=O
138.13
138.13
Crystalline Powder
315°C
C6H6N2O2
1 g
NMCKJFCJIHCHIS-UHFFFAOYSA-N
6-aminopyridine-2-carboxylic acid
351652
≥98.0% (HPLC,T)
6-Amino-2-pyridinecarboxylic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'