Learn More
6-Amino-5-bromoquinoline, 97%, Thermo Scientific Chemicals
$337.85 - $1132.23
Chemical Identifiers
CAS | 50358-42-4 |
---|---|
Molecular Formula | C9H7BrN2 |
Molecular Weight (g/mol) | 223.073 |
MDL Number | MFCD00234503 |
InChI Key | MODLGTLYXJGDCH-UHFFFAOYSA-N |
PubChem CID | 12741232 |
IUPAC Name | 5-bromoquinolin-6-amine |
SMILES | C1=CC2=C(C=CC(=C2Br)N)N=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6315003
|
Thermo Scientific Chemicals
H6315003 |
1 g |
Each for $337.85
|
|
||||||
AAH6315006
|
Thermo Scientific Chemicals
H6315006 |
5 g |
Each for $1,132.23
|
|
||||||
Description
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.
Solubility
Slightly soluble in water. Soluble in alcohol, ether, and carbon disulfide.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
50358-42-4 | |
223.073 | |
MODLGTLYXJGDCH-UHFFFAOYSA-N | |
5-bromoquinolin-6-amine |
C9H7BrN2 | |
MFCD00234503 | |
12741232 | |
C1=CC2=C(C=CC(=C2Br)N)N=C1 |
Specifications
50358-42-4 | |
C9H7BrN2 | |
1 g | |
Slightly soluble in water. Soluble in alcohol,ether,and carbon disulfide. | |
C1=CC2=C(C=CC(=C2Br)N)N=C1 | |
223.073 | |
223.07 | |
Solid |
Yellow to Brown | |
MFCD00234503 | |
6-amino-5-bromoquinoline, 6-quinolinamine, 5-bromo, 5-bromo-6-quinolinamine, 6-quinolinamine,5-bromo, 5-bromo-quinolin-6-ylamine, 6-quinolinamine, 5-bromo-9ci | |
MODLGTLYXJGDCH-UHFFFAOYSA-N | |
5-bromoquinolin-6-amine | |
12741232 | |
97% | |
6-Amino-5-bromoquinoline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only