missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Amino-5-nitro-2-picoline, 98%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 360780010

Catalog No. AC360780010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

 

Specifications

Specifications

6-Amino-5-nitro-2-picoline
97.5
Authentic
Glass bottle
MFCD00047443
2-amino-3-nitro-6-picoline, 2-amino-6-methyl-3-nitropyridine, 2-amino-3-nitro-6-methylpyridine, 6-amino-5-nitro-2-picoline, 6-methyl-3-nitro-pyridin-2-ylamine, 6-methyl-3-nitro-2-pyridinamine, 2-pyridinamine, 6-methyl-3-nitro, 6-methyl-3-nitro-2-pyridylamine, 6-methyl-3-nitropyridin-2-amin, 2-amino-3-nitro-6-picoline 2-amino-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])N
153
153
21901-29-1
100.0
97.5% min. (GC)
C6H7N3O2
1g
LCJXSRQGDONHRK-UHFFFAOYSA-N
6-methyl-3-nitropyridin-2-amine
226028
98%

Chemical Identifiers

21901-29-1
153
LCJXSRQGDONHRK-UHFFFAOYSA-N
226028
CC1=NC(=C(C=C1)[N+](=O)[O-])N
C6H7N3O2
MFCD00047443
2-amino-3-nitro-6-picoline, 2-amino-6-methyl-3-nitropyridine, 2-amino-3-nitro-6-methylpyridine, 6-amino-5-nitro-2-picoline, 6-methyl-3-nitro-pyridin-2-ylamine, 6-methyl-3-nitro-2-pyridinamine, 2-pyridinamine, 6-methyl-3-nitro, 6-methyl-3-nitro-2-pyridylamine, 6-methyl-3-nitropyridin-2-amin, 2-amino-3-nitro-6-picoline 2-amino-6-methyl-3-nitropyridine
6-methyl-3-nitropyridin-2-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only