missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Aminocaproic acid, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 103301000

 View more versions of this product

Catalog No. AC103301000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Aminocaproic acid
204.0°C to 209.0°C
Authentic
Plastic bottle
H2N(CH2)5CO2H
04,434
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
SLXKOJJOQWFEFD-UHFFFAOYSA-N
6-aminohexanoic acid
564
131.17
Crystalline Powder
60-32-2
White
99+%
C6H13NO2
100 g
15,427
(13g/100ml H2O) Clear colorless solution
C(CCC(=O)O)CCN
131.17
CHEBI:16586
99+%

Chemical Identifiers

60-32-2
131.17
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
CHEBI:16586
C(CCC(=O)O)CCN
C6H13NO2
SLXKOJJOQWFEFD-UHFFFAOYSA-N
564
6-aminohexanoic acid
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 200-469-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only