Learn More
6-Aminoquinoline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 369640010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
6-Aminoquinoline | |
115.0°C to 119.0°C | |
97.5% min. (HPLC) | |
C9H8N2 | |
22, V,10, 303 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, benzene, diethyl ether,heptane | |
C1=CC2=C(C=CC(=C2)N)N=C1 | |
144.18 | |
144.18 |
580-15-4 | |
192.0°C to 195.0°C (14.0 mmHg) | |
Glass bottle | |
1 g | |
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine | |
RJSRSRITMWVIQT-UHFFFAOYSA-N | |
quinolin-6-amine | |
11373 | |
98% |
Chemical Identifiers
580-15-4 | |
144.18 | |
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine | |
quinolin-6-amine |
C9H8N2 | |
RJSRSRITMWVIQT-UHFFFAOYSA-N | |
11373 | |
C1=CC2=C(C=CC(=C2)N)N=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 209-453-
RUO – Research Use Only