missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Bromo-[1,2,4]triazolo[1,5-a]pyridine, 96%, Thermo Scientific Chemicals
$75.25 - $206.36
Chemical Identifiers
CAS | 356560-80-0 |
---|---|
Molecular Formula | C6H4BrN3 |
Molecular Weight (g/mol) | 198.023 |
MDL Number | MFCD08689534 |
InChI Key | CXRXKDSDRWLKTK-UHFFFAOYSA-N |
Synonym | 6-bromo-1,2,4 triazolo 1,5-a pyridine, 6-bromo 1,2,4 triazolo 1,5-a pyridine, 1,2,4 triazolo 1,5-a pyridine, 6-bromo, 6-bromo-1,2,4-triazolo-1,5-a-pyridine, 6-bromo-1,2,4-triazolo 1,5-a pyridine, 1,2,4 triazolo 1,5-a pyridine,6-bromo, 6-bromanyl-1,2,4 triazolo 1,5-a pyridine, 6-bromo 1,2,4 triazolo 1,5-alpha pyridine, tert-butyl 6e-7-4-4-fluorophenyl-6-isopropyl-2-methyl methylsulfonyl amino pyrimidin-5-yl-3r,5s-3,5-dihydroxyhept-6-enoate |
PubChem CID | 11513934 |
IUPAC Name | 6-bromo-[1,2,4]triazolo[1,5-a]pyridine |
SMILES | C1=CC2=NC=NN2C=C1Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH66700MD
|
Thermo Scientific Chemicals
H66700MD |
250 mg |
N/A
|
|
||||||
AAH6670003
|
Thermo Scientific Chemicals
H6670003 |
1 g |
N/A
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
356560-80-0 | |
198.023 | |
CXRXKDSDRWLKTK-UHFFFAOYSA-N | |
11513934 | |
C1=CC2=NC=NN2C=C1Br |
C6H4BrN3 | |
MFCD08689534 | |
6-bromo-1,2,4 triazolo 1,5-a pyridine, 6-bromo 1,2,4 triazolo 1,5-a pyridine, 1,2,4 triazolo 1,5-a pyridine, 6-bromo, 6-bromo-1,2,4-triazolo-1,5-a-pyridine, 6-bromo-1,2,4-triazolo 1,5-a pyridine, 1,2,4 triazolo 1,5-a pyridine,6-bromo, 6-bromanyl-1,2,4 triazolo 1,5-a pyridine, 6-bromo 1,2,4 triazolo 1,5-alpha pyridine, tert-butyl 6e-7-4-4-fluorophenyl-6-isopropyl-2-methyl methylsulfonyl amino pyrimidin-5-yl-3r,5s-3,5-dihydroxyhept-6-enoate | |
6-bromo-[1,2,4]triazolo[1,5-a]pyridine |
Specifications
356560-80-0 | |
MFCD08689534 | |
10061274 | |
CXRXKDSDRWLKTK-UHFFFAOYSA-N | |
6-bromo-[1,2,4]triazolo[1,5-a]pyridine | |
11513934 | |
96% | |
6-Bromo-[1,2,4]triazolo[1,5-a]pyridine |
C6H4BrN3 | |
250 mg | |
6-bromo-1,2,4 triazolo 1,5-a pyridine, 6-bromo 1,2,4 triazolo 1,5-a pyridine, 1,2,4 triazolo 1,5-a pyridine, 6-bromo, 6-bromo-1,2,4-triazolo-1,5-a-pyridine, 6-bromo-1,2,4-triazolo 1,5-a pyridine, 1,2,4 triazolo 1,5-a pyridine,6-bromo, 6-bromanyl-1,2,4 triazolo 1,5-a pyridine, 6-bromo 1,2,4 triazolo 1,5-alpha pyridine, tert-butyl 6e-7-4-4-fluorophenyl-6-isopropyl-2-methyl methylsulfonyl amino pyrimidin-5-yl-3r,5s-3,5-dihydroxyhept-6-enoate | |
C1=CC2=NC=NN2C=C1Br | |
198.023 | |
198.02 | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only