missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-1-hexene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 364700010

 View more versions of this product

Catalog No. AC364700010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Bromo-1-hexene
96.0
Yellow
47.0°C to 51.0°C (16.0 mmHg)
Authentic
Glass bottle
1.4645 to 1.4665 (20°C, 589nm)
MFCD00000269
1.217
RIMXEJYJXDBLIE-UHFFFAOYSA-N
6-bromohex-1-ene
75906
97%
2695-47-8
100.0
1.2170g/mL
52°C
96% min. (GC)
C6H11Br
Br(CH2)4CH=CH2
1 g
6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene
C=CCCCCBr
163.06
163.06
Liquid

Chemical Identifiers

2695-47-8
163.06
RIMXEJYJXDBLIE-UHFFFAOYSA-N
75906
C=CCCCCBr
C6H11Br
MFCD00000269
6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene
6-bromohex-1-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

EINECSNumber : 220-267-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only