missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromo-2,4-dimethylpyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2746003

 View more versions of this product

Catalog No. AAH2746003



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Bromo-2,4-dimethylpyridine-3-boronic acid
∽145°C (decomposition)
MFCD09037491
6-bromo-2,4-dimethylpyridine-3-boronic acid, 6-bromo-2,4-dimethylpyridin-3-yl boronic acid, acmc-2098oh, 6-bromo-2,4-dimethylpyridin-3-yl boronicacid
B(C1=C(N=C(C=C1C)Br)C)(O)O
229.868
229.87
1072944-23-0
C7H9BBrNO2
1 g
GXXITYQLWOMIEM-UHFFFAOYSA-N
(6-bromo-2,4-dimethylpyridin-3-yl)boronic acid
44755186
95%

Chemical Identifiers

1072944-23-0
229.868
GXXITYQLWOMIEM-UHFFFAOYSA-N
44755186
B(C1=C(N=C(C=C1C)Br)C)(O)O
C7H9BBrNO2
MFCD09037491
6-bromo-2,4-dimethylpyridine-3-boronic acid, 6-bromo-2,4-dimethylpyridin-3-yl boronic acid, acmc-2098oh, 6-bromo-2,4-dimethylpyridin-3-yl boronicacid
(6-bromo-2,4-dimethylpyridin-3-yl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only