missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-hydroxycoumarin 98.0+%, TCI America™

$80.16 - $240.46

Chemical Identifiers

CAS 4139-61-1
Molecular Formula C9H5BrO3
Molecular Weight (g/mol) 241.04
MDL Number MFCD00239372
InChI Key KNMCTCABMSGXGR-UHFFFAOYSA-N
PubChem CID 54691404
IUPAC Name 6-bromo-4-hydroxychromen-2-one
SMILES C1=CC2=C(C=C1Br)C(=CC(=O)O2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B46991G
View Documents
TCI America
B46991G
1 g
Each for $80.16
Only null left
 
B46995G
View Documents
TCI America
B46995G
5 g
Each for $240.46
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4139-61-1
241.04
KNMCTCABMSGXGR-UHFFFAOYSA-N
6-bromo-4-hydroxychromen-2-one
C9H5BrO3
MFCD00239372
54691404
C1=CC2=C(C=C1Br)C(=CC(=O)O2)O

Specifications

4139-61-1
C9H5BrO3
1 g
C1=CC2=C(C=C1Br)C(=CC(=O)O2)O
241.04
241.04
Crystalline Powder
277°C
MFCD00239372
KNMCTCABMSGXGR-UHFFFAOYSA-N
6-bromo-4-hydroxychromen-2-one
54691404
≥98.0% (GC,T)
6-Bromo-4-hydroxycoumarin
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'