missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromohexanoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 165670250

 View more versions of this product

Catalog No. AC165670250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Bromohexanoic acid
32.0°C to 35.0°C
165.0°C to 170.0°C (20.0 mmHg)
Authentic
Glass bottle
Br(CH2)5CO2H
02,II,287
NVRVNSHHLPQGCU-UHFFFAOYSA-N
6-bromohexanoic acid
20210
195.06
Low Melting Crystals
4224-70-8
Orange to White
67°C
98%
C6H11BrO2
25 g
6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid
C(CCC(=O)O)CCBr
195.06
CHEBI:60700
98%

Chemical Identifiers

4224-70-8
195.06
6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid
CHEBI:60700
C(CCC(=O)O)CCBr
C6H11BrO2
NVRVNSHHLPQGCU-UHFFFAOYSA-N
20210
6-bromohexanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 224-176-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only