Learn More
6-Bromohexanoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 165671000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
6-Bromohexanoic acid | |
32.0°C to 35.0°C | |
165.0°C to 170.0°C (20.0 mmHg) | |
Authentic | |
Glass bottle | |
Br(CH2)5CO2H | |
02,II,287 | |
NVRVNSHHLPQGCU-UHFFFAOYSA-N | |
6-bromohexanoic acid | |
20210 | |
195.06 | |
Low Melting Crystals |
4224-70-8 | |
Orange to White | |
67°C | |
98% | |
C6H11BrO2 | |
100 g | |
6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid | |
C(CCC(=O)O)CCBr | |
195.06 | |
CHEBI:60700 | |
98% |
Chemical Identifiers
4224-70-8 | |
195.06 | |
6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid | |
CHEBI:60700 | |
C(CCC(=O)O)CCBr |
C6H11BrO2 | |
NVRVNSHHLPQGCU-UHFFFAOYSA-N | |
20210 | |
6-bromohexanoic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 224-176-5
RUO – Research Use Only