missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromoindole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H63799MD

 View more versions of this product

Catalog No. AAH63799MD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Bromoindole-2-carboxylic acid
C9H6BrNO2
250 mg
SVBVYRYROZWKNJ-UHFFFAOYSA-N
6-bromo-1H-indole-2-carboxylic acid
4011696
97%
16732-65-3
MFCD02664469
6-bromoindole-2-carboxylic acid, 6-bromo-2-indolecarboxylic acid, 1h-indole-2-carboxylic acid, 6-bromo, 6-bromoindole-2-carboxylicacid, zlchem 475, pubchem1664, acmc-1bqkh, intermediates-zcf02255, evoblocks eb21020, timtec-bb sbb014280
C1=CC2=C(C=C1Br)NC(=C2)C(=O)O
240.056
240.05
Solid

Chemical Identifiers

16732-65-3
240.056
SVBVYRYROZWKNJ-UHFFFAOYSA-N
4011696
C1=CC2=C(C=C1Br)NC(=C2)C(=O)O
C9H6BrNO2
MFCD02664469
6-bromoindole-2-carboxylic acid, 6-bromo-2-indolecarboxylic acid, 1h-indole-2-carboxylic acid, 6-bromo, 6-bromoindole-2-carboxylicacid, zlchem 475, pubchem1664, acmc-1bqkh, intermediates-zcf02255, evoblocks eb21020, timtec-bb sbb014280
6-bromo-1H-indole-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only