missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-1-hexyne, 97%

CAS: 10297-06-0 | C6H9Cl | 116.59 g/mol

$115.70 - $407.86

Chemical Identifiers

CAS 10297-06-0
Molecular Formula C6H9Cl
Molecular Weight (g/mol) 116.59
MDL Number MFCD00013697
InChI Key ZUKOCGMVJUXIJA-UHFFFAOYSA-N
PubChem CID 272001
IUPAC Name 6-chlorohex-1-yne
SMILES ClCCCCC#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC380810050
SDS View Documents
Thermo Scientific Chemicals
380810050
5 mL Glass bottle
Each for $115.70
Only null left
 
AC380810250
SDS View Documents
Thermo Scientific Chemicals
380810250
25 mL Glass bottle
Each for $407.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10297-06-0
116.59
ZUKOCGMVJUXIJA-UHFFFAOYSA-N
6-chlorohex-1-yne
C6H9Cl
MFCD00013697
272001
ClCCCCC#C

Specifications

10297-06-0
100.0
0.9620g/mL
37°C
96% min. (GC)
C6H9Cl
CH∽C(CH2)4Cl
5 mL
6-chloro-1-hexyne, 5-hexynechloride, 1-hexyne,6-chloro, 1-hexyne, 6-chloro, 5-hexynyl chloride, 1-chloro-5-hexyne, 6-chloro-hex-1-yne, acmc-1boe0, zukocgmvjuxija-uhfffaoysa
ClCCCCC#C
116.59
116.59
Liquid
96.0
Yellow
131°C
Authentic
Glass bottle
1.4490 to 1.451
MFCD00013697
0.962
ZUKOCGMVJUXIJA-UHFFFAOYSA-N
6-chlorohex-1-yne
272001
97%
6-Chloro-1-hexyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only