missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Chloro-1-hydroxy-1H-benzotriazole, 97%, Thermo Scientific™
$152.54 - $152.54
Chemical Identifiers
CAS | 26198-19-6 |
---|---|
Molecular Formula | C6H4ClN3O |
Molecular Weight (g/mol) | 169.57 |
InChI Key | TZCYLJGNWDVJRA-UHFFFAOYSA-N |
Synonym | cl-hobt, 6-chloro-1h-benzo d 1,2,3 triazol-1-ol, 6-chloro-1-hydroxybenzotriazol, 6-chloro-1-hydroxy-1h-benzotriazole, 6-chloro-1-hydroxibenzotriazol, 6-chloro-1h-benzotriazol-1-ol, 6-chloro-1h-1,2,3-benzotriazol-1-ol, 1-hydroxy-6-chlorobenzotriazole, 6-chloro-1,2,3-benzotriazol-1-ol, 1h-benzotriazole, 6-chloro-1-hydroxy |
PubChem CID | 232711 |
IUPAC Name | 6-chloro-1-hydroxybenzotriazole |
SMILES | C1=CC2=C(C=C1Cl)N(N=N2)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC448920250
|
Thermo Scientific Chemicals
448920250 |
25 g |
Each for $152.54
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
26198-19-6 | |
169.57 | |
cl-hobt, 6-chloro-1h-benzo d 1,2,3 triazol-1-ol, 6-chloro-1-hydroxybenzotriazol, 6-chloro-1-hydroxy-1h-benzotriazole, 6-chloro-1-hydroxibenzotriazol, 6-chloro-1h-benzotriazol-1-ol, 6-chloro-1h-1,2,3-benzotriazol-1-ol, 1-hydroxy-6-chlorobenzotriazole, 6-chloro-1,2,3-benzotriazol-1-ol, 1h-benzotriazole, 6-chloro-1-hydroxy | |
6-chloro-1-hydroxybenzotriazole |
C6H4ClN3O | |
TZCYLJGNWDVJRA-UHFFFAOYSA-N | |
232711 | |
C1=CC2=C(C=C1Cl)N(N=N2)O |
Specifications
26198-19-6 | |
96% min. (HPLC) | |
C6H4ClN3O | |
cl-hobt, 6-chloro-1h-benzo d 1,2,3 triazol-1-ol, 6-chloro-1-hydroxybenzotriazol, 6-chloro-1-hydroxy-1h-benzotriazole, 6-chloro-1-hydroxibenzotriazol, 6-chloro-1h-benzotriazol-1-ol, 6-chloro-1h-1,2,3-benzotriazol-1-ol, 1-hydroxy-6-chlorobenzotriazole, 6-chloro-1,2,3-benzotriazol-1-ol, 1h-benzotriazole, 6-chloro-1-hydroxy | |
C1=CC2=C(C=C1Cl)N(N=N2)O | |
169.57 | |
169.57 | |
6-Chloro-1-hydroxy-1H-benzotriazole |
190.0°C to 195.0°C | |
Glass bottle | |
25 g | |
TZCYLJGNWDVJRA-UHFFFAOYSA-N | |
6-chloro-1-hydroxybenzotriazole | |
232711 | |
97% |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only