missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-2-(1,4-diazepan-1-yl)-1,3-benzothiazole, 95+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AW00326DA

Catalog No. AW00326DA


Explore available promotions

Description

Description

Specifications

Specifications

6-Chloro-2-(1, 4-diazepan-1-yl)-1, 3-benzothiazole
94.5
95+%
C12H14ClN3S
1 g
UZLJYDCBJULBQU-UHFFFAOYSA-N
6-chloro-2-(1,4-diazepan-1-yl)-1,3-benzothiazole
2795675
95%
348134-09-8
100.0
Amber Glass Bottle
MFCD00661816
6-chloro-2-1,4-diazepan-1-yl-1,3-benzothiazole, maybridge1_000355, 6-chloro-2-1,4-diazepane-1-yl-1,3-benzothiazole, 6-chloro-2-1,4 diazepan-1-yl-benzothiazole, 6-chloro-2-1.4 diazepan-1-yl-benzothiazole, 6-chloro-2-1,4-diazepan-1-yl benzo d thiazole
C1CNCCN(C1)C2=NC3=C(S2)C=C(C=C3)Cl
267.775
267.78

Chemical Identifiers

348134-09-8
267.775
UZLJYDCBJULBQU-UHFFFAOYSA-N
2795675
C1CNCCN(C1)C2=NC3=C(S2)C=C(C=C3)Cl
C12H14ClN3S
MFCD00661816
6-chloro-2-1,4-diazepan-1-yl-1,3-benzothiazole, maybridge1_000355, 6-chloro-2-1,4-diazepane-1-yl-1,3-benzothiazole, 6-chloro-2-1,4 diazepan-1-yl-benzothiazole, 6-chloro-2-1.4 diazepan-1-yl-benzothiazole, 6-chloro-2-1,4-diazepan-1-yl benzo d thiazole
6-chloro-2-(1,4-diazepan-1-yl)-1,3-benzothiazole
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions