missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals

$121.81 - $121.81

Chemical Identifiers

CAS 19064-67-6
Molecular Formula C4H3ClN2O
Molecular Weight (g/mol) 130.531
MDL Number MFCD00160460
InChI Key YICPBKWYZXFJNB-UHFFFAOYSA-N
Synonym 6-chloropyridazin-3-ol, 6-chloro-3-hydroxypyridazine, 6-chloro-2h-pyridazin-3-one, 6-chloropyridazin-3 2h-one, 3-chloro-6-hydroxypyridazine, 3 2h-pyridazinone, 6-chloro, 6-chloro-3 2h-pyridazinone, 3-chloro-6-pyridazone, 3-hydroxy-6-chloropyridazine, 6-chloro-3-pyridazinol
PubChem CID 252828
IUPAC Name 3-chloro-1H-pyridazin-6-one
SMILES C1=CC(=NNC1=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6183706
View Documents
Thermo Scientific Chemicals
H6183706
5 g
Each for $121.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19064-67-6
130.531
YICPBKWYZXFJNB-UHFFFAOYSA-N
252828
C1=CC(=NNC1=O)Cl
C4H3ClN2O
MFCD00160460
6-chloropyridazin-3-ol, 6-chloro-3-hydroxypyridazine, 6-chloro-2h-pyridazin-3-one, 6-chloropyridazin-3 2h-one, 3-chloro-6-hydroxypyridazine, 3 2h-pyridazinone, 6-chloro, 6-chloro-3 2h-pyridazinone, 3-chloro-6-pyridazone, 3-hydroxy-6-chloropyridazine, 6-chloro-3-pyridazinol
3-chloro-1H-pyridazin-6-one

Specifications

19064-67-6
MFCD00160460
6-chloropyridazin-3-ol, 6-chloro-3-hydroxypyridazine, 6-chloro-2h-pyridazin-3-one, 6-chloropyridazin-3 2h-one, 3-chloro-6-hydroxypyridazine, 3 2h-pyridazinone, 6-chloro, 6-chloro-3 2h-pyridazinone, 3-chloro-6-pyridazone, 3-hydroxy-6-chloropyridazine, 6-chloro-3-pyridazinol
C1=CC(=NNC1=O)Cl
130.531
130.53
6-Chloro-3(2H)-pyridazinone
C4H3ClN2O
5 g
YICPBKWYZXFJNB-UHFFFAOYSA-N
3-chloro-1H-pyridazin-6-one
252828
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only