missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-chloroacetyl-2-benzoxazolinone, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 439050010

 View more versions of this product

Catalog No. AC439050010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

6-chloroacetyl-2-benzoxazolinone
96.0
Authentic
Glass Bottle
1g
KOOWFDRPKXUCRF-UHFFFAOYSA-N
6-(2-chloroacetyl)-3H-1,3-benzoxazol-2-one
1988981
97%
54903-10-5
100.0
96% min. (HPLC)
C9H6ClNO3
6-chloroacetyl-2-benzoxazolinone, jpb 24, 2 3h-benzoxazolone, 6-chloroacetyl, 6-2-chloroacetyl benzo d oxazol-2 3h-one, 6-chloroacetyl-2 3h-benzoxazolone, 6-2-chloroacetyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-2-chloroacetyl-3-hydrobenzoxazol-2-one, 6-2-chloroacetyl-3h-1,3-benzoxazol-2-one, acmc-20alz8, 6-chloroacetyl benzoxazole-2 3h-one
C1=CC2=C(C=C1C(=O)CCl)OC(=O)N2
211.6
211.6

Chemical Identifiers

54903-10-5
211.6
6-chloroacetyl-2-benzoxazolinone, jpb 24, 2 3h-benzoxazolone, 6-chloroacetyl, 6-2-chloroacetyl benzo d oxazol-2 3h-one, 6-chloroacetyl-2 3h-benzoxazolone, 6-2-chloroacetyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-2-chloroacetyl-3-hydrobenzoxazol-2-one, 6-2-chloroacetyl-3h-1,3-benzoxazol-2-one, acmc-20alz8, 6-chloroacetyl benzoxazole-2 3h-one
6-(2-chloroacetyl)-3H-1,3-benzoxazol-2-one
C9H6ClNO3
KOOWFDRPKXUCRF-UHFFFAOYSA-N
1988981
C1=CC2=C(C=C1C(=O)CCl)OC(=O)N2
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only