missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Chlorochroman-4-one, 99%, Thermo Scientific Chemicals
$85.41 - $1063.21
Chemical Identifiers
CAS | 37674-72-9 |
---|---|
Molecular Formula | C9H7ClO2 |
Molecular Weight (g/mol) | 182.603 |
MDL Number | MFCD00024070 |
InChI Key | LLTDYHFVIVSQPJ-UHFFFAOYSA-N |
Synonym | 6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5 |
PubChem CID | 95273 |
IUPAC Name | 6-chloro-2,3-dihydrochromen-4-one |
SMILES | C1COC2=C(C1=O)C=C(C=C2)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1497503
|
Thermo Scientific Chemicals
A1497503 |
1 g |
Each for $85.41
|
|
|||||
AAA1497506
|
Thermo Scientific Chemicals
A1497506 |
5 g |
Each for $262.78
|
|
|||||
AAA1497514
|
Thermo Scientific Chemicals
A1497514 |
25 g |
Each for $1,063.21
|
|
|||||
Description
6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.
Solubility
Soluble in solvents.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
37674-72-9 | |
182.603 | |
LLTDYHFVIVSQPJ-UHFFFAOYSA-N | |
95273 | |
C1COC2=C(C1=O)C=C(C=C2)Cl |
C9H7ClO2 | |
MFCD00024070 | |
6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5 | |
6-chloro-2,3-dihydrochromen-4-one |
Specifications
37674-72-9 | |
C9H7ClO2 | |
1 g | |
6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5 | |
LLTDYHFVIVSQPJ-UHFFFAOYSA-N | |
6-chloro-2,3-dihydrochromen-4-one | |
95273 | |
99% |
101°C to 102°C | |
MFCD00024070 | |
124218 | |
Soluble in solvents. | |
C1COC2=C(C1=O)C=C(C=C2)Cl | |
182.603 | |
182.61 | |
6-Chlorochroman-4-one |
Safety and Handling
EINECSNumber : 253-611-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only