missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorohexanoic Acid 98.0+%, TCI America™

$134.21 - $397.05

Chemical Identifiers

CAS 4224-62-8
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00061103
InChI Key XWWKSLXUVZVGSP-UHFFFAOYSA-N
PubChem CID 20209
IUPAC Name 6-chlorohexanoic acid
SMILES OC(=O)CCCCCCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C15075G
View Documents
TCI America
C15075G
5 g N/A
Only null left
 
C150725G
View Documents
TCI America
C150725G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4224-62-8
150.60
XWWKSLXUVZVGSP-UHFFFAOYSA-N
6-chlorohexanoic acid
C6H11ClO2
MFCD00061103
20209
OC(=O)CCCCCCl

Specifications

4224-62-8
C6H11ClO2
5 g
XWWKSLXUVZVGSP-UHFFFAOYSA-N
6-chlorohexanoic acid
20209
≥98.0% (GC,T)
6-Chlorohexanoic Acid
White-Yellow
MFCD00061103
3261
OC(=O)CCCCCCl
150.60
150.60
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1180

RTECSNumber : MO7000000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'