missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole-2-carboxylic acid, 96%, Thermo Scientific Chemicals

$122.33 - $423.67

Chemical Identifiers

CAS 16732-75-5
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.602
MDL Number MFCD01863157
InChI Key BKPSJOSKWKTWAG-UHFFFAOYSA-N
Synonym 6-chloroindole-2-carboxylic acid, pubchem1671, acmc-209dwc, chembl23134, 6-chloro-2-indolecarboxylic acid, 6-chloro indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-chloro, 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-2-indolecarboxylic acid
PubChem CID 85584
IUPAC Name 6-chloro-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Cl)NC(=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6686403
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
H6686403
1 g
Each for $122.33
Only null left
 
AAH6686406
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
H6686406
5 g
Each for $423.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16732-75-5
195.602
BKPSJOSKWKTWAG-UHFFFAOYSA-N
85584
C1=CC2=C(C=C1Cl)NC(=C2)C(=O)O
C9H6ClNO2
MFCD01863157
6-chloroindole-2-carboxylic acid, pubchem1671, acmc-209dwc, chembl23134, 6-chloro-2-indolecarboxylic acid, 6-chloro indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-chloro, 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-2-indolecarboxylic acid
6-chloro-1H-indole-2-carboxylic acid

Specifications

16732-75-5
MFCD01863157
152660
BKPSJOSKWKTWAG-UHFFFAOYSA-N
6-chloro-1H-indole-2-carboxylic acid
85584
96%
6-Chloroindole-2-carboxylic acid
C9H6ClNO2
1 g
6-chloroindole-2-carboxylic acid, pubchem1671, acmc-209dwc, chembl23134, 6-chloro-2-indolecarboxylic acid, 6-chloro indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-chloro, 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-2-indolecarboxylic acid
C1=CC2=C(C=C1Cl)NC(=C2)C(=O)O
195.602
195.6
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 240-799-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only