missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chlorothiochroman-4-one, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1030406

 View more versions of this product

Catalog No. AAA1030406


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Chlorothiochroman-4-one
65°C to 69°C
MFCD00052997
124621
OKHUUKHZUNKSQA-UHFFFAOYSA-N
6-chloro-2,3-dihydrothiochromen-4-one
248065
98%
13735-12-1
C9H7ClOS
5 g
6-chlorothiochroman-4-one, 6-chloro-2,3-dihydro-4h-thiochromen-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiopyran-4-one, 6-chloro-thiochroman-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiin-4-one, 6-chloro-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-chloro-2,3-dihydro-1-benzothiopyran-4-one, 6-chloro-2h,3h-benzo e thiin-4-one, acmc-20alz1, maybridge1_007431
C1CSC2=C(C1=O)C=C(C=C2)Cl
198.664
198.67

Chemical Identifiers

13735-12-1
198.664
OKHUUKHZUNKSQA-UHFFFAOYSA-N
248065
C1CSC2=C(C1=O)C=C(C=C2)Cl
C9H7ClOS
MFCD00052997
6-chlorothiochroman-4-one, 6-chloro-2,3-dihydro-4h-thiochromen-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiopyran-4-one, 6-chloro-thiochroman-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiin-4-one, 6-chloro-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-chloro-2,3-dihydro-1-benzothiopyran-4-one, 6-chloro-2h,3h-benzo e thiin-4-one, acmc-20alz1, maybridge1_007431
6-chloro-2,3-dihydrothiochromen-4-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only