missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
$126.81 - $421.36
Chemical Identifiers
CAS | 947191-69-7 |
---|---|
Molecular Formula | C15H22BNO3 |
Molecular Weight (g/mol) | 275.155 |
MDL Number | MFCD07781184 |
InChI Key | QEQNYJUPYICBCR-UHFFFAOYSA-N |
Synonym | 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-cyclopropylmethoxy pyridine-3-boronic acid pinacol ester, 2-cyclopropylmethoxy pyridine-5-boronic acid, pinacol ester, 6-cyclopropylmethoxypyridine-3-boronic acid pinacol ester, 6-cyclopropylmethoxypyridine-3-yl boronic acid pinacol ester, 6-cyclopropylmethoxy pyridin-3-yl boronic acid pinacol ester, 2-cyclopropyl-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine |
PubChem CID | 24208797 |
IUPAC Name | 2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3CC3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH50139MD
|
Thermo Scientific Chemicals
H50139MD |
250 mg |
Each for $126.81
|
|
|||||
AAH5013903
|
Thermo Scientific Chemicals
H5013903 |
1 g |
Each for $421.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
947191-69-7 | |
275.155 | |
QEQNYJUPYICBCR-UHFFFAOYSA-N | |
24208797 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3CC3 |
C15H22BNO3 | |
MFCD07781184 | |
2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-cyclopropylmethoxy pyridine-3-boronic acid pinacol ester, 2-cyclopropylmethoxy pyridine-5-boronic acid, pinacol ester, 6-cyclopropylmethoxypyridine-3-boronic acid pinacol ester, 6-cyclopropylmethoxypyridine-3-yl boronic acid pinacol ester, 6-cyclopropylmethoxy pyridin-3-yl boronic acid pinacol ester, 2-cyclopropyl-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine | |
2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Specifications
947191-69-7 | |
MFCD07781184 | |
Air Sensitive | |
QEQNYJUPYICBCR-UHFFFAOYSA-N | |
2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
24208797 | |
97% |
C15H22BNO3 | |
250 mg | |
2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-cyclopropylmethoxy pyridine-3-boronic acid pinacol ester, 2-cyclopropylmethoxy pyridine-5-boronic acid, pinacol ester, 6-cyclopropylmethoxypyridine-3-boronic acid pinacol ester, 6-cyclopropylmethoxypyridine-3-yl boronic acid pinacol ester, 6-cyclopropylmethoxy pyridin-3-yl boronic acid pinacol ester, 2-cyclopropyl-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3CC3 | |
275.155 | |
275.16 | |
6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only