missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals 6-Fluoro-DL-tryptophan, 97%
Supplier: Thermo Scientific Chemicals 146741000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
6-Fluoro-DL-tryptophan | |
96.0 | |
280°C to 285°C | |
Authentic | |
Glass bottle | |
MFCD00005653 | |
6-fluoro-dl-tryptophan, 6-fluorotryptophan, 2-amino-3-6-fluoro-1h-indol-3-yl propanoic acid, dl-tryptophan, 6-fluoro, dl-6-fluorotryptophane, +--6-fluorotryptophan, tryptophan, 6-fluoro-, dl, 6-fluoro-d,l-tryptophan, tryptophan, 6-fluoro, 2-amino-3-6-fluoroindol-3-yl propanoic acid | |
NC(CC1=CNC2=CC(F)=CC=C12)C(O)=O | |
222.22 | |
222.22 | |
Powder |
7730-20-3 | |
100.0 | |
Beige to White | |
97% | |
C11H11FN2O2 | |
100 mg | |
YMEXGEAJNZRQEH-UHFFFAOYSA-N | |
2-amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid | |
94937 | |
97% |
Chemical Identifiers
7730-20-3 | |
222.22 | |
YMEXGEAJNZRQEH-UHFFFAOYSA-N | |
94937 | |
NC(CC1=CNC2=CC(F)=CC=C12)C(O)=O |
C11H11FN2O2 | |
MFCD00005653 | |
6-fluoro-dl-tryptophan, 6-fluorotryptophan, 2-amino-3-6-fluoro-1h-indol-3-yl propanoic acid, dl-tryptophan, 6-fluoro, dl-6-fluorotryptophane, +--6-fluorotryptophan, tryptophan, 6-fluoro-, dl, 6-fluoro-d,l-tryptophan, tryptophan, 6-fluoro, 2-amino-3-6-fluoroindol-3-yl propanoic acid | |
2-amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid |
Safety and Handling
EINECSNumber : 231-788-6
RUO – Research Use Only