missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(Fmoc-amino)hexanoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6314806

 View more versions of this product

Catalog No. AAH6314806


Explore available promotions

Description

Description

6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1% acetic acid, water and 1mmol in 2mL DMF clearly soluble.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

6-(Fmoc-amino)hexanoic acid
White
FmocNH(CH2)5CO2H
5 g
fmoc-6-ahx-oh, fmoc-epsilon-acp-oh, n-fmoc-6-aminohexanoic acid, fmoc-, a-acp-oh, 6-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxycarbonylamino hexanoic acid, hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino
FPCPONSZWYDXRD-UHFFFAOYSA-N
6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
2756087
98%
88574-06-5
C21H23NO4
MFCD00077045
5883220
Soluble in 1% acetic acid,water and 1mmol in 2mL DMF clearly soluble.
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCC(=O)O
353.418
353.41
Powder

Chemical Identifiers

88574-06-5
353.418
FPCPONSZWYDXRD-UHFFFAOYSA-N
2756087
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCC(=O)O
C21H23NO4
MFCD00077045
fmoc-6-ahx-oh, fmoc-epsilon-acp-oh, n-fmoc-6-aminohexanoic acid, fmoc-, a-acp-oh, 6-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxycarbonylamino hexanoic acid, hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino
6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only