missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Mercapto-1-hexanol 98.0+%, TCI America™
Supplier: TCI America M226625G
Specifications
6-Mercapto-1-hexanol | |
Colorless | |
C6H14OS | |
25 g | |
C(CCCS)CCO | |
134.237 | |
134.24 | |
Liquid |
1633-78-9 | |
105°C | |
MFCD00068552 | |
UGZAJZLUKVKCBM-UHFFFAOYSA-N | |
6-sulfanylhexan-1-ol | |
560126 | |
≥98.0% (GC) |
Chemical Identifiers
1633-78-9 | |
134.237 | |
UGZAJZLUKVKCBM-UHFFFAOYSA-N | |
6-sulfanylhexan-1-ol |
C6H14OS | |
MFCD00068552 | |
560126 | |
C(CCCS)CCO |
Safety and Handling
TSCA : No