Learn More
6-Methoxyquinoline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 179340250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
6-Methoxyquinoline | |
5263-87-6 | |
100.0 | |
White to Cream | |
193.0°C (50.0 mmHg) | |
Authentic | |
Glass bottle | |
1.6170 to 1.6240 | |
25 g | |
1.152 | |
Solubility in water: insoluble | |
COC1=CC2=C(C=C1)N=CC=C2 | |
159.19 | |
CHEBI:72822 | |
98% |
98% | |
97.5 | |
18.0°C to 20.0°C | |
1.1520g/mL | |
>110°C | |
97.5% min. (GC) | |
C10H9NO | |
MFCD00006800 | |
21,85 | |
quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q | |
HFDLDPJYCIEXJP-UHFFFAOYSA-N | |
6-methoxyquinoline | |
14860 | |
159.19 | |
Granular Powder |
Chemical Identifiers
5263-87-6 | |
159.19 | |
HFDLDPJYCIEXJP-UHFFFAOYSA-N | |
14860 | |
6-methoxyquinoline |
C10H9NO | |
MFCD00006800 | |
quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q | |
CHEBI:72822 | |
COC1=CC2=C(C=C1)N=CC=C2 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water f
GHS Signal Word: Warning
EINECSNumber : 226-077-2
TSCA : TSCA
RUO – Research Use Only