missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methyl-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals

$275.75 - $830.92

Chemical Identifiers

CAS 13327-27-0
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00039720
InChI Key QZWIXLPWMGHDDD-UHFFFAOYSA-N
Synonym 6-methylpyridazin-3 2h-one, 6-methyl-3 2h-pyridazinone, 6-methylpyridazin-3-ol, 6-methyl-2h-pyridazin-3-one, 6-methyl-3-pyridazinone, 3-hydroxy-6-methylpyridazine, 3 2h-pyridazinone, 6-methyl, 6-methyl-2,3-dihydropyridazin-3-one, 6me3oxopyridaz
PubChem CID 83346
IUPAC Name 3-methyl-1H-pyridazin-6-one
SMILES CC1=NNC(=O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1447106
View Documents
Thermo Scientific Chemicals
A1447106
5 g
Each for $275.75
 
AAA1447114
View Documents
Thermo Scientific Chemicals
A1447114
25 g
Each for $830.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13327-27-0
110.12
QZWIXLPWMGHDDD-UHFFFAOYSA-N
83346
CC1=NNC(=O)C=C1
C5H6N2O
MFCD00039720
6-methylpyridazin-3 2h-one, 6-methyl-3 2h-pyridazinone, 6-methylpyridazin-3-ol, 6-methyl-2h-pyridazin-3-one, 6-methyl-3-pyridazinone, 3-hydroxy-6-methylpyridazine, 3 2h-pyridazinone, 6-methyl, 6-methyl-2,3-dihydropyridazin-3-one, 6me3oxopyridaz
3-methyl-1H-pyridazin-6-one

Specifications

13327-27-0
C5H6N2O
5 g
6-methylpyridazin-3 2h-one, 6-methyl-3 2h-pyridazinone, 6-methylpyridazin-3-ol, 6-methyl-2h-pyridazin-3-one, 6-methyl-3-pyridazinone, 3-hydroxy-6-methylpyridazine, 3 2h-pyridazinone, 6-methyl, 6-methyl-2,3-dihydropyridazin-3-one, 6me3oxopyridaz
CC1=NNC(=O)C=C1
110.12
110.12
6-Methyl-3(2H)-pyridazinone
143°C to 145°C
MFCD00039720
109928
QZWIXLPWMGHDDD-UHFFFAOYSA-N
3-methyl-1H-pyridazin-6-one
83346
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-367-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only