missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methyl-5-hepten-2-one, 98%, Thermo Scientific Chemicals

$46.60 - $62.45

Chemical Identifiers

CAS 110-93-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00008905
InChI Key UHEPJGULSIKKTP-UHFFFAOYSA-N
Synonym 6-methyl-5-hepten-2-one, sulcatone, methylheptenone, 5-hepten-2-one, 6-methyl, 2-methyl-2-hepten-6-one, methyl heptenone, 6-methyl-5-heptene-2-one, heptenone, methyl, 2-methyl-6-oxo-2-heptene, 2-oxo-6-methylhept-5-ene
PubChem CID 9862
ChEBI CHEBI:16310
IUPAC Name 6-methylhept-5-en-2-one
SMILES CC(C)=CCCC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1333514
View Documents
Thermo Scientific Chemicals
A1333514
25 g
Each for $46.60
 
AAA1333522
View Documents
Thermo Scientific Chemicals
A1333522
100 g
Each for $62.45
 
Description

Description

6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

Solubility
Soluble in methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container.
Specifications

Chemical Identifiers

110-93-0
126.20
UHEPJGULSIKKTP-UHFFFAOYSA-N
9862
6-methylhept-5-en-2-one
C8H14O
MFCD00008905
6-methyl-5-hepten-2-one, sulcatone, methylheptenone, 5-hepten-2-one, 6-methyl, 2-methyl-2-hepten-6-one, methyl heptenone, 6-methyl-5-heptene-2-one, heptenone, methyl, 2-methyl-6-oxo-2-heptene, 2-oxo-6-methylhept-5-ene
CHEBI:16310
CC(C)=CCCC(C)=O

Specifications

110-93-0
0.855
56°C (133°F)
1.439
25 g
1741705
Soluble in methanol and chloroform.
CC(C)=CCCC(C)=O
126.20
CHEBI:16310
98%
-67°C
172°C to 174°C
C8H14O
MFCD00008905
UN1224
6-methyl-5-hepten-2-one, sulcatone, methylheptenone, 5-hepten-2-one, 6-methyl, 2-methyl-2-hepten-6-one, methyl heptenone, 6-methyl-5-heptene-2-one, heptenone, methyl, 2-methyl-6-oxo-2-heptene, 2-oxo-6-methylhept-5-ene
UHEPJGULSIKKTP-UHFFFAOYSA-N
6-methylhept-5-en-2-one
9862
126.2
6-Methyl-5-hepten-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c

H226-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 203-816-7

RTECSNumber : MJ9700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only