missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methylindole, 99%, Thermo Scientific Chemicals

$343.43 - $343.43

Chemical Identifiers

CAS 3420-02-8
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00005682
InChI Key ONYNOPPOVKYGRS-UHFFFAOYSA-N
Synonym n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester
PubChem CID 137930
SMILES CC1=CC=C2C=CNC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC133880010
SDS View Documents
Thermo Scientific Chemicals
133880010
1 g Glass bottle
Each for $343.43
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3420-02-8
131.18
ONYNOPPOVKYGRS-UHFFFAOYSA-N
137930
C9H9N
MFCD00005682
n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester
CC1=CC=C2C=CNC2=C1

Specifications

3420-02-8
100.0
1.0500g/mL
>112°C
98.5% min. (GC)
C9H9N
MFCD00005682
20, IV, 3214
n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester
CC1=CC=C2C=CNC2=C1
131.18
131.18
Liquid After Melting
98.5
Yellow to Brown
112°C (5.0 mmHg)
Authentic
Glass bottle
1.6060 to 1.608
1 g
1.05
ONYNOPPOVKYGRS-UHFFFAOYSA-N
tert-butyl N-phenylcarbamate
137930
99%
6-Methylindole
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : This product darkens on exposure to light

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only