Learn More
6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
$449.48 - $1023.48
Chemical Identifiers
CAS | 1032758-87-4 |
---|---|
Molecular Formula | C17H27BN2O4 |
Molecular Weight (g/mol) | 334.223 |
MDL Number | MFCD09037484 |
InChI Key | QGBJKHALDLKAGX-UHFFFAOYSA-N |
Synonym | tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-boc-methylamino pyridine-3-boronic acid pinacol ester, 6-n-boc-methylamino pyridine-3-boronic acid pinacol ester, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-tert-butoxycarbonyl methyl amino pyridin-3-yl boronic acid pinacol ester, tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-yl carbamate, tert-butylmethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate |
PubChem CID | 44541676 |
IUPAC Name | tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)N(C)C(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH27430MD
|
Thermo Scientific Chemicals
H27430MD |
250 mg |
Each for $449.48
|
|
|||||
AAH2743003
|
Thermo Scientific Chemicals
H2743003 |
1 g |
Each for $1,023.48
|
|
|||||
Description
Used in the synthesis of organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the synthesis of organic compounds.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
1032758-87-4 | |
334.223 | |
QGBJKHALDLKAGX-UHFFFAOYSA-N | |
44541676 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)N(C)C(=O)OC(C)(C)C |
C17H27BN2O4 | |
MFCD09037484 | |
tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-boc-methylamino pyridine-3-boronic acid pinacol ester, 6-n-boc-methylamino pyridine-3-boronic acid pinacol ester, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-tert-butoxycarbonyl methyl amino pyridin-3-yl boronic acid pinacol ester, tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-yl carbamate, tert-butylmethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate | |
tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate |
Specifications
1032758-87-4 | |
C17H27BN2O4 | |
250 mg | |
QGBJKHALDLKAGX-UHFFFAOYSA-N | |
tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate | |
44541676 | |
95% |
112°C to 119°C | |
MFCD09037484 | |
tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-boc-methylamino pyridine-3-boronic acid pinacol ester, 6-n-boc-methylamino pyridine-3-boronic acid pinacol ester, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-tert-butoxycarbonyl methyl amino pyridin-3-yl boronic acid pinacol ester, tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-yl carbamate, tert-butylmethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)N(C)C(=O)OC(C)(C)C | |
334.223 | |
334.22 | |
6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only