missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Nitropiperonyl alcohol, 98+%, Thermo Scientific Chemicals
$221.23 - $914.62
Chemical Identifiers
CAS | 15341-08-9 |
---|---|
Molecular Formula | C8H7NO5 |
Molecular Weight (g/mol) | 197.15 |
MDL Number | MFCD00005825 |
InChI Key | LJXBRPHBDWIHPR-UHFFFAOYSA-N |
Synonym | 6-nitropiperonyl alcohol, 6-nitrobenzo d 1,3 dioxol-5-yl methanol, 3,4-methylenedioxy-6-nitrobenzyl alcohol, 6-nitro-3,4-methylenedioxybenzyl alcohol, 6-nitro-1,3-benzodioxol-5-yl methanol, 6-nitro-2h-1,3-benzodioxol-5-yl methanol, 6-nitro-1,3-benzodioxole-5-methanol, 1,3-benzodioxole-5-methanol,6-nitro, 1,3-benzodioxole-5-methanol, 6-nitro, 6-nitro-1,3-benzodioxol-5-yl methanol # |
PubChem CID | 519111 |
IUPAC Name | (5-nitro-2H-1,3-benzodioxol-4-yl)methanol |
SMILES | OCC1=C2OCOC2=CC=C1[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1814006
|
Thermo Scientific Chemicals
A1814006 |
5 g |
Each for $221.23
|
|
|||||
AAA1814014
|
Thermo Scientific Chemicals
A1814014 |
25 g |
Each for $914.62
|
|
|||||
Description
6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.
Notes
Light sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
15341-08-9 | |
197.15 | |
LJXBRPHBDWIHPR-UHFFFAOYSA-N | |
519111 | |
OCC1=C2OCOC2=CC=C1[N+]([O-])=O |
C8H7NO5 | |
MFCD00005825 | |
6-nitropiperonyl alcohol, 6-nitrobenzo d 1,3 dioxol-5-yl methanol, 3,4-methylenedioxy-6-nitrobenzyl alcohol, 6-nitro-3,4-methylenedioxybenzyl alcohol, 6-nitro-1,3-benzodioxol-5-yl methanol, 6-nitro-2h-1,3-benzodioxol-5-yl methanol, 6-nitro-1,3-benzodioxole-5-methanol, 1,3-benzodioxole-5-methanol,6-nitro, 1,3-benzodioxole-5-methanol, 6-nitro, 6-nitro-1,3-benzodioxol-5-yl methanol # | |
(5-nitro-2H-1,3-benzodioxol-4-yl)methanol |
Specifications
15341-08-9 | |
C8H7NO5 | |
5 g | |
Light sensitive | |
LJXBRPHBDWIHPR-UHFFFAOYSA-N | |
(5-nitro-2H-1,3-benzodioxol-4-yl)methanol | |
519111 | |
≥98% |
121°C to 125°C | |
MFCD00005825 | |
13749 | |
6-nitropiperonyl alcohol, 6-nitrobenzo d 1,3 dioxol-5-yl methanol, 3,4-methylenedioxy-6-nitrobenzyl alcohol, 6-nitro-3,4-methylenedioxybenzyl alcohol, 6-nitro-1,3-benzodioxol-5-yl methanol, 6-nitro-2h-1,3-benzodioxol-5-yl methanol, 6-nitro-1,3-benzodioxole-5-methanol, 1,3-benzodioxole-5-methanol,6-nitro, 1,3-benzodioxole-5-methanol, 6-nitro, 6-nitro-1,3-benzodioxol-5-yl methanol # | |
OCC1=C2OCOC2=CC=C1[N+]([O-])=O | |
197.15 | |
197.15 | |
6-Nitropiperonyl alcohol |
Safety and Handling
EINECSNumber : 239-376-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only