missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Phenoxynicotinaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 173282-69-4
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD02681950
InChI Key SOPOQXNWJNVZMI-UHFFFAOYSA-N
Synonym 6-phenoxynicotinaldehyde, 6-phenoxypyridine-3-carboxaldehyde, 2-phenoxypyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde,6-phenoxy, 5-formyl-2-phenoxypyridine, 6-phenoxy-3-pyridinecarboxaldehyde, 6-phenoxy-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 6-phenoxy
PubChem CID 2776500
IUPAC Name 6-phenoxypyridine-3-carbaldehyde
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC19604CB
SDS View Documents
Thermo Scientific Chemicals
CC19604CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC19604DA
SDS View Documents
Thermo Scientific Chemicals
CC19604DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC19604DE
SDS View Documents
Thermo Scientific Chemicals
CC19604DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

173282-69-4
199.209
SOPOQXNWJNVZMI-UHFFFAOYSA-N
2776500
C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O
C12H9NO2
MFCD02681950
6-phenoxynicotinaldehyde, 6-phenoxypyridine-3-carboxaldehyde, 2-phenoxypyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde,6-phenoxy, 5-formyl-2-phenoxypyridine, 6-phenoxy-3-pyridinecarboxaldehyde, 6-phenoxy-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 6-phenoxy
6-phenoxypyridine-3-carbaldehyde

Specifications

173282-69-4
100.0
Amber Glass Bottle
MFCD02681950
6-phenoxynicotinaldehyde, 6-phenoxypyridine-3-carboxaldehyde, 2-phenoxypyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde,6-phenoxy, 5-formyl-2-phenoxypyridine, 6-phenoxy-3-pyridinecarboxaldehyde, 6-phenoxy-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 6-phenoxy
C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O
199.209
199.21
6-Phenoxynicotinaldehyde
96.5
97%
C12H9NO2
250mg
SOPOQXNWJNVZMI-UHFFFAOYSA-N
6-phenoxypyridine-3-carbaldehyde
2776500
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'