Learn More
6-(Tetrahydropyran-4-yloxy)nicotinic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC58301CB
Specifications
6-(Tetrahydropyran-4-yloxy)nicotinic acid | |
96.5 | |
97% | |
C11H13NO4 | |
250 mg | |
OLOSQIMMFCSJHD-UHFFFAOYSA-N | |
6-(oxan-4-yloxy)pyridine-3-carboxylic acid | |
24229576 | |
97% |
886851-55-4 | |
100.0 | |
Amber Glass Bottle | |
MFCD09258833 | |
6-oxan-4-yloxy pyridine-3-carboxylic acid, 6-tetrahydropyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yl oxy nicotinic acid, 2-tetrahydro-2h-pyran-4-yloxy pyridine-5-carboxylic acid, 3-pyridinecarboxylicacid, 6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yl oxy pyridine-3-carboxylic acid, 4-5-carboxypyridin-2-yl oxy tetrahydro-2h-pyran | |
C1COCCC1OC2=NC=C(C=C2)C(=O)O | |
223.228 | |
223.23 |
Chemical Identifiers
886851-55-4 | |
223.228 | |
OLOSQIMMFCSJHD-UHFFFAOYSA-N | |
24229576 | |
C1COCCC1OC2=NC=C(C=C2)C(=O)O |
C11H13NO4 | |
MFCD09258833 | |
6-oxan-4-yloxy pyridine-3-carboxylic acid, 6-tetrahydropyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yl oxy nicotinic acid, 2-tetrahydro-2h-pyran-4-yloxy pyridine-5-carboxylic acid, 3-pyridinecarboxylicacid, 6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yl oxy pyridine-3-carboxylic acid, 4-5-carboxypyridin-2-yl oxy tetrahydro-2h-pyran | |
6-(oxan-4-yloxy)pyridine-3-carboxylic acid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning