missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(Tetrahydropyran-4-yloxy)nicotinonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC58316DE

 View more versions of this product

Catalog No. CC58316DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

6-(Tetrahydropyran-4-yloxy)nicotinonitrile
96.5
97%
C11H12N2O2
5g
CCEWHGWDICBASG-UHFFFAOYSA-N
6-(oxan-4-yloxy)pyridine-3-carbonitrile
24229580
97%
884507-60-2
100.0
Amber Glass Bottle
MFCD09702388
6-tetrahydropyran-4-yloxy nicotinonitrile, 6-tetrahydro-2h-pyran-4-yloxy pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yloxy pyridine-3-carbonitrile, 6-oxan-4-yl oxy pyridine-3-carbonitrile, 6-tetrahydro-2h-pyran-4-yl oxy nicotinonitrile, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-3-carbonitrile
N#CC1=CN=C(OC2CCOCC2)C=C1
204.23
204.23

Chemical Identifiers

884507-60-2
204.23
CCEWHGWDICBASG-UHFFFAOYSA-N
24229580
N#CC1=CN=C(OC2CCOCC2)C=C1
C11H12N2O2
MFCD09702388
6-tetrahydropyran-4-yloxy nicotinonitrile, 6-tetrahydro-2h-pyran-4-yloxy pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yloxy pyridine-3-carbonitrile, 6-oxan-4-yl oxy pyridine-3-carbonitrile, 6-tetrahydro-2h-pyran-4-yl oxy nicotinonitrile, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-3-carbonitrile
6-(oxan-4-yloxy)pyridine-3-carbonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions