missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Thioguanine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2128003

 View more versions of this product

Catalog No. AAB2128003


Explore available promotions

Description

Description

6-Thioguanine acts as an antineoplastic and purine antimetabolite. It is also useful as an inhibition of stimulated expression of TNF-related apoptosis-inducing ligand (TRAIL) protein. It is involved in the treatment of acute leukemias and Psoriasis. Further, it is used for the treatment of ulcerative colitis and autoimmune diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

6-Thioguanine
>360°C
C5H5N5S
1 g
157765
6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol
WYWHKKSPHMUBEB-UHFFFAOYSA-N
2-amino-3,7-dihydropurine-6-thione
2723601
167.19
154-42-7
0.98
MFCD00233553
UN2811
14,9337
Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.
NC1=NC(=S)C2=C(N1)N=CN2
167.19
CHEBI:9555
98%

Chemical Identifiers

154-42-7
167.19
WYWHKKSPHMUBEB-UHFFFAOYSA-N
2723601
NC1=NC(=S)C2=C(N1)N=CN2
C5H5N5S
MFCD00233553
6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol
CHEBI:9555
Safety and Handling

Safety and Handling

GHS H Statement
H301

P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c

H301-H341

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 205-827-2

RTECSNumber : UP0740000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only