Learn More
6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals
$65.64 - $205.82
Chemical Identifiers
CAS | 131747-42-7 |
---|---|
Molecular Formula | C7H4F3NO2 |
Molecular Weight (g/mol) | 191.11 |
MDL Number | MFCD02180819 |
InChI Key | OKBHXGBLXDNJJD-UHFFFAOYSA-N |
Synonym | 6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g |
PubChem CID | 14761453 |
IUPAC Name | 6-(trifluoromethyl)pyridine-2-carboxylic acid |
SMILES | OC(=O)C1=NC(=CC=C1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63138MD
|
Thermo Scientific Chemicals
H63138MD |
250 mg |
Each for $65.64
|
|
|||||
AAH6313803
|
Thermo Scientific Chemicals
H6313803 |
1 g |
Each for $205.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
131747-42-7 | |
191.11 | |
OKBHXGBLXDNJJD-UHFFFAOYSA-N | |
14761453 | |
OC(=O)C1=NC(=CC=C1)C(F)(F)F |
C7H4F3NO2 | |
MFCD02180819 | |
6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g | |
6-(trifluoromethyl)pyridine-2-carboxylic acid |
Specifications
131747-42-7 | |
White | |
MFCD02180819 | |
UN2811 | |
OKBHXGBLXDNJJD-UHFFFAOYSA-N | |
6-(trifluoromethyl)pyridine-2-carboxylic acid | |
14761453 | |
98% | |
6-(Trifluoromethyl)pyridine-2-carboxylic acid |
154°C to 158°C | |
C7H4F3NO2 | |
250 mg | |
6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g | |
OC(=O)C1=NC(=CC=C1)C(F)(F)F | |
191.11 | |
191.11 | |
Solid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only