missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(Trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H3403303

Catalog No. AAH3403303


Explore available promotions

Description

Description

6-Trifluoromethyl-3-pyridinesulfonyl Chloride is a chemical used for the preparation of N-sulfonamido polycyclic pyrazolyl compounds for treatment of cognitive disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Trifluoromethyl-3-pyridinesulfonyl Chloride is a chemical used for the preparation of N-sulfonamido polycyclic pyrazolyl compounds for treatment of cognitive disorders.

Notes
Store in cool place. Keep container ti
Specifications

Specifications

6-(Trifluoromethyl)pyridine-3-sulfonyl chloride
42°C to 45°C
MFCD10699112
UN3261
NSGLNIQAWVNZFB-UHFFFAOYSA-N
6-(trifluoromethyl)pyridine-3-sulfonyl chloride
45594311
97%
959996-58-8
C6H3ClF3NO2S
1 g
6-trifluoromethyl pyridine-3-sulfonyl chloride, 6-trifluoromethyl-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride,6-trifluoromethyl, chloro 6-trifluoromethyl 3-pyridyl sulfone, 3-pyridinesulfonyl chloride, 6-trifluoromethyl
C1=CC(=NC=C1S(=O)(=O)Cl)C(F)(F)F
245.6
245.61

Chemical Identifiers

959996-58-8
245.6
NSGLNIQAWVNZFB-UHFFFAOYSA-N
45594311
C1=CC(=NC=C1S(=O)(=O)Cl)C(F)(F)F
C6H3ClF3NO2S
MFCD10699112
6-trifluoromethyl pyridine-3-sulfonyl chloride, 6-trifluoromethyl-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride,6-trifluoromethyl, chloro 6-trifluoromethyl 3-pyridyl sulfone, 3-pyridinesulfonyl chloride, 6-trifluoromethyl
6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only