missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laxco™ 60° Brix Calibration Fluid for Handheld Analog Refractometers
Used to Calibrate Handheld Analog Brix Refractometers
Supplier: Laxco™ RPCAL60P
Comprend: 5ml droplet bottle of brix 60° solution
Description
- 5mL droplet bottle of 60° Brix Calibration solution for calibration of Handheld Analog Refractometer models for brix models range from 45° to 90°
- Quantity: 3
Spécifications
| Calibration Fluid | |
| Refractometer | |
| ±0.1° | |
| 5 years optical and mechanical, 1 year electrical from date of purchase |
| 60 Brix | |
| 5mL bottle | |
| 5 Pk. |