3M™ 6000 Series Half Face Mask Respirator Edge
Edge
Half-face mask respirators 6000 Series are economical, low-maintenance, and extremely lightweight. Probed reusable respirator assembly 6000Q series are used for quantitative fit purposes only.
Supplier:  3M™ 7000001932
 View more versions of this product
Catalog No. 17-986-9A
Edge
Explore available promotions
Description
Description
Combines features of both reusable and disposable respirators to provide effective protection. 3M™ Half Face Mask Respirator 6000 Series are economical, low-maintenance, simple-to-handle, and extremely lightweight.
  • Reusable half masks are economical, low-maintenance, simple-to-handle, and extremely lightweight
  • Used with 3M™ Supplied Air Systems
  • Bayonet Connection System allows connection to a broad range of twin lightweight filters to protect against gases, vapors, and particulates
  • Twin filter design provides lower breathing resistance
  • Probed reusable respirator assembly 6000 Series used for quantitative fit testing (17-987-3A & -3B)
Specifications
Specifications
EN 140:1998
Bayonet
Thermoplastic Elastomer
Small
Cartridge 6000 Series, Filter 2000, 2200, 7000 or 5000 Series, or Dual Airline System
6100
NIOSH 42 CFR Part 84
Gray
Half-Mask Respirator
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
Reclosable storage bag
4-point
SDS
Documents
Documents
Accessories