Learn More
Takara Bio Usa 6X HIS MONOCLONAL ANTIBOD DFS Item
Supplier:  Clontech Laboratories 631212
6xHis Monoclonal Antibody (Albumin Free), 200microgram, Detect all 6xHis-tagged proteins, IgG2a isotype,
Catalog No. NC9720312
Only null left
Explore available promotions