Learn More
7,8-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals
$29.96 - $95.70
Chemical Identifiers
CAS | 2107-77-9 |
---|---|
Molecular Formula | C10H8O4 |
Molecular Weight (g/mol) | 192.17 |
MDL Number | MFCD00016972 |
InChI Key | NWQBYMPNIJXFNQ-UHFFFAOYSA-N |
Synonym | 7,8-dihydroxy-4-methylcoumarin, 4-methyldaphnetin, 7,8-dihydroxy-4-methyl-2h-chromen-2-one, 4-methyldaphnetine, 4-methyl-7,8-dihydroxycoumarin, 2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl, coumarin, 7,8-dihydroxy-4-methyl, 7,8-dihydroxy-4-methyl-2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl-chromen-2-one, 7,8-dihydroxy-4-methyl-2-benzopyrone |
PubChem CID | 5355836 |
SMILES | CC1=CC(=O)OC2=C(O)C(O)=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL14069MC
|
Thermo Scientific Chemicals
L14069MC |
100 mg |
Each for $29.96
|
|
||||||
AAL14069ME
|
Thermo Scientific Chemicals
L14069ME |
500 mg |
Each for $63.92
|
|
||||||
AAL1406903
|
Thermo Scientific Chemicals
L1406903 |
1 g |
Each for $95.70
|
|
||||||
Description
Can be used as a fluorescent sensor to monitor the consumption of a boronic acid in a Suzuki cross-coupling reaction. Use a long wave UV lamp.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2107-77-9 | |
192.17 | |
NWQBYMPNIJXFNQ-UHFFFAOYSA-N | |
5355836 |
C10H8O4 | |
MFCD00016972 | |
7,8-dihydroxy-4-methylcoumarin, 4-methyldaphnetin, 7,8-dihydroxy-4-methyl-2h-chromen-2-one, 4-methyldaphnetine, 4-methyl-7,8-dihydroxycoumarin, 2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl, coumarin, 7,8-dihydroxy-4-methyl, 7,8-dihydroxy-4-methyl-2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl-chromen-2-one, 7,8-dihydroxy-4-methyl-2-benzopyrone | |
CC1=CC(=O)OC2=C(O)C(O)=CC=C12 |
Specifications
2107-77-9 | |
C10H8O4 | |
100 mg | |
Light sensitive | |
NWQBYMPNIJXFNQ-UHFFFAOYSA-N | |
7,8-dihydroxy-4-methyl-2H-chromen-2-one | |
5355836 | |
97% |
241°C to 244°C | |
MFCD00016972 | |
177613 | |
7,8-dihydroxy-4-methylcoumarin, 4-methyldaphnetin, 7,8-dihydroxy-4-methyl-2h-chromen-2-one, 4-methyldaphnetine, 4-methyl-7,8-dihydroxycoumarin, 2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl, coumarin, 7,8-dihydroxy-4-methyl, 7,8-dihydroxy-4-methyl-2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl-chromen-2-one, 7,8-dihydroxy-4-methyl-2-benzopyrone | |
CC1=CC(=O)OC2=C(O)C(O)=CC=C12 | |
192.17 | |
192.17 | |
7,8-Dihydroxy-4-methylcoumarin |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-290-4
RTECSNumber : GN6385000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only